Gene Transcripts
- Gene Symbol
- Mir489
- Entrez Gene ID
- 100314071
- Gene Name
- microRNA mir-489
- Gene Aliases
- rno-mir-489
- Chromosome Location
- Chr. 4: 28659523 - 28659628 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_032105.1 |
|
1 |
87 |
|
View Full Product Details