Gene Transcripts

Gene Symbol
Mgarp
Entrez Gene ID
67749
Gene Name
mitochondria localized glutamic acid rich protein
Gene Aliases
4930583H14Rik, AI195347, CESP-1, HUMMR, Osap, Qsap
Chromosome Location
Chr. 3: 51388412 - 51396547 on Build GRCm38
UniGene ID
Mm.273339
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028121.1 1 230
GenBank mRNA BC022740.1 - 1 185

View Full Product Details