Gene Transcripts
- Gene Symbol
- Gm11346
- Entrez Gene ID
- 76024
- Gene Name
- X-linked lymphocyte-regulated 5 pseudogene
- Gene Aliases
- 5830431A10Rik, AI447617, Gm11347, OTTMUSG00000000601, OTTMUSG00000000602
- Chromosome Location
- Chr. 13: 24598124 - 24604767 on Build GRCm38
- UniGene ID
- Mm.440710
- Species
- Mus musculus
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CJ236201.1 |
- |
1 |
125 |
|
Chr. 13 - 24598030 - 24598693 [-] |
View Full Product Details