Gene Transcripts
- Gene Symbol
- Mir489
- Entrez Gene ID
- 723877
- Gene Name
- microRNA 489
- Gene Aliases
- Mirn489, mir-489, mmu-mir-489
- Chromosome Location
- Chr. 6: 3721897 - 3722003 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_030250.1 |
|
1 |
94 |
|
View Full Product Details