Gene Transcripts
- Gene Symbol
- 2610001J05Rik
- Entrez Gene ID
- 66520
- Gene Name
- RIKEN cDNA 2610001J05 gene
- Gene Aliases
- C79299
- Chromosome Location
- Chr. 6: 13869074 - 13871483 on Build GRCm38
- UniGene ID
- Mm.29615
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024619.1 |
|
3 |
1368 |
|
GenBank mRNA |
AK035027.1 |
- |
3 |
1351 |
|
AK152937.1 |
- |
|
1404 |
|
View Full Product Details