Gene Transcripts
- Gene Symbol
- Gm2518
- Entrez Gene ID
- 100039957
- Gene Name
- interferon induced transmembrane protein 2 pseudogene
- Gene Aliases
- D630022E15Rik
- Chromosome Location
- Chr. 19: 8774485 - 8776962 on Build GRCm38
- UniGene ID
- Mm.220886
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_015538.1 |
|
1 |
2289 |
|
GenBank mRNA |
AK082962.1 |
- |
1 |
2288 |
|
View Full Product Details