Gene Transcripts

Gene Symbol
Gm2518
Entrez Gene ID
100039957
Gene Name
interferon induced transmembrane protein 2 pseudogene
Gene Aliases
D630022E15Rik
Chromosome Location
Chr. 19: 8774485 - 8776962 on Build GRCm38
UniGene ID
Mm.220886
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015538.1 1 894
GenBank mRNA AK041085.1 - 2 790
AK077736.1 - 1 891
AK082962.1 - 1 893
AK085400.1 - 1 866

View Full Product Details