Gene Transcripts
- Gene Symbol
- Gm10638
- Entrez Gene ID
- 666945
- Gene Name
- predicted gene 10638
- Gene Aliases
- ENSMUSG00000074178
- Chromosome Location
- Chr. 8: 86745699 - 86747060 on Build GRCm38
- UniGene ID
- Mm.391806
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027829.1 |
|
1 |
568 |
|
GenBank mRNA |
AK133050.1 |
- |
1 |
569 |
|
AK133595.1 |
- |
|
94 |
|
View Full Product Details