Gene Transcripts
- Gene Symbol
- Stoml1
- Entrez Gene ID
- 69106
- Gene Name
- stomatin-like 1
- Gene Aliases
- 1810015E19Rik, AI847431, SLP1, STORP, UNC24, WPB72
- Chromosome Location
- Chr. 9: 58253164 - 58262524 on Build GRCm38
- UniGene ID
- Mm.216502
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028146.1 |
|
3 |
518 |
|
GenBank mRNA |
AK189780.1 |
- |
2 |
195 |
|
BC017536.1 |
- |
3 |
490 |
|
View Full Product Details