Gene Transcripts

Gene Symbol
Stoml1
Entrez Gene ID
69106
Gene Name
stomatin-like 1
Gene Aliases
1810015E19Rik, AI847431, SLP1, STORP, UNC24, WPB72
Chromosome Location
Chr. 9: 58253164 - 58262524 on Build GRCm38
UniGene ID
Mm.216502
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028146.1 3 434
GenBank mRNA AK189780.1 - 2 111
BC017536.1 - 3 406

View Full Product Details