Gene Transcripts
- Gene Symbol
- 1700084E18Rik
- Entrez Gene ID
- 67350
- Gene Name
- RIKEN cDNA 1700084E18 gene
- Gene Aliases
- Chromosome Location
- Chr. 2: 30237197 - 30237631 on Build GRCm38
- UniGene ID
- Mm.402683
- Species
- Mus musculus
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY707421.1 |
- |
1 |
266 |
|
Chr. 2 - 30237191 - 30237519 [-] |
GenBankmRNA |
BB090636.2 |
- |
1 |
200 |
|
Chr. 2 - 30236799 - 30237454 [-] |
GenBankmRNA |
BY707046.1 |
- |
1 |
378 |
|
Chr. 2 - 30237200 - 30237631 [-] |
GenBankmRNA |
BE137757.1 |
- |
1 |
353 |
|
Chr. 2 - 30237215 - 30237607 [-] |
GenBankmRNA |
BY707385.1 |
- |
1 |
266 |
|
Chr. 2 - 30237191 - 30237519 [-] |
View Full Product Details