Gene Transcripts

Gene Symbol
5033406O09Rik
Entrez Gene ID
77675
Gene Name
RIKEN cDNA 5033406O09 gene
Gene Aliases
9130204G15Rik
Chromosome Location
Chr. 12: 111941991 - 111944482 on Build GRCm38
UniGene ID
Mm.98331
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_029464.1 2 437
GenBank mRNA AK017159.1 - 2 397
AK020272.1 - 2 164
AK079861.1 - 2 431
AK134157.1 - 2 371

View Full Product Details