Gene Transcripts
- Gene Symbol
- 5033406O09Rik
- Entrez Gene ID
- 77675
- Gene Name
- RIKEN cDNA 5033406O09 gene
- Gene Aliases
- 9130204G15Rik
- Chromosome Location
- Chr. 12: 111941991 - 111944482 on Build GRCm38
- UniGene ID
- Mm.98331
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_029464.1 |
|
3 |
1267 |
|
GenBank mRNA |
AK020272.1 |
- |
3 |
994 |
|
AK134157.1 |
- |
3 |
1203 |
|
View Full Product Details