Gene Transcripts

Gene Symbol
BMS1P4
Entrez Gene ID
729096
Gene Name
BMS1, ribosome biogenesis factor pseudogene 4
Gene Aliases
BMS1LP4
Chromosome Location
Chr. 10: 73699151 - 73730514 on Build GRCh38
UniGene ID
Hs.709171
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026592.2 10 1399

Gene Symbol
BMS1P5
Entrez Gene ID
399761
Gene Name
BMS1, ribosome biogenesis factor pseudogene 5
Gene Aliases
BMS1LP1, BMS1LP5, BMS1P1, bA556L1.3
Chromosome Location
Chr. 10: 46786733 - 46811989 on Build GRCh38
UniGene ID
Hs.711898
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AL832069.1 - 8 3815

Gene Symbol
BMS1P6
Entrez Gene ID
642826
Gene Name
BMS1, ribosome biogenesis factor pseudogene 6
Gene Aliases
BMS1LP2, BMS1LP6, BMS1P2, bA302K17.2
Chromosome Location
Chr. 10: 47535574 - 47553523 on Build GRCh38
UniGene ID
Hs.463017
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024495.2 9 1209
GenBank mRNA AK094691.1 - 1 539
BC051298.1 - 8 1175


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BI962861.1 - 1 276 Chr. 10 - 73704225 - 73713509 [-]
GenBankmRNA CN431124.1 - 1 508 Chr. 10 - 73704951 - 73713715 [-]
GenBankmRNA BG993955.1 - 2 270 Chr. 10 - 73705020 - 73718166 [-]
GenBankmRNA CF995158.1 - 4 512 Chr. 10 - 73704579 - 73720632 [-]

View Full Product Details