Gene Transcripts

Gene Symbol
FAM86HP
Entrez Gene ID
729375
Gene Name
family with sequence similarity 86, member A pseudogene
Gene Aliases
Chromosome Location
Chr. 3: 130097782 - 130111433 on Build GRCh38
UniGene ID
Hs.123633
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024252.1 2 177
GenBank mRNA DA296336.1 - 2 177


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AK125025.1 - 1 2544 Chr. 3 - 130104681 - 130108152 [-]

View Full Product Details