Gene Transcripts

Gene Symbol
FAM86HP
Entrez Gene ID
729375
Gene Name
family with sequence similarity 86, member A pseudogene
Gene Aliases
Chromosome Location
Chr. 3: 130097782 - 130111433 on Build GRCh38
UniGene ID
Hs.123633
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024252.1 4 485
GenBank mRNA AK124448.1 - 2 255
DA296336.1 - 4 485

View Full Product Details