Gene Transcripts

Gene Symbol
CST13P
Entrez Gene ID
164380
Gene Name
cystatin 13, pseudogene
Gene Aliases
CSTT, CTES6
Chromosome Location
Chr. 20: 23519146 - 23542018 on Build GRCh38
UniGene ID
Hs.400007
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_001279.2 2 559
GenBank mRNA BC024006.1 - 2 564
BC028368.1 - 2 564
BC033934.1 - 2 554
BC041171.1 - 2 538

View Full Product Details