Gene Transcripts

Gene Symbol
CLLU1
Entrez Gene ID
574028
Gene Name
chronic lymphocytic leukemia up-regulated 1
Gene Aliases
Chromosome Location
Chr. 12: 92421531 - 92431002 on Build GRCh38
UniGene ID
Hs.730377
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027932.1 1 74
NR_027933.1 1 74
GenBank mRNA AJ845162.1 - 1 74
AJ845163.1 - 1 74
AJ845164.1 - 1 74
AJ845167.1 - 1 74

View Full Product Details