Gene Transcripts
- Gene Symbol
- MIR490
- Entrez Gene ID
- 574443
- Gene Name
- microRNA 490
- Gene Aliases
- MIRN490, hsa-mir-490, miR-490
- Chromosome Location
- Chr. 7: 136903167 - 136903294 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_030165.1 |
|
1 |
60 |
|
View Full Product Details