Gene Transcripts
- Gene Symbol
- LOC148696
- Entrez Gene ID
- 148696
- Gene Name
- uncharacterized LOC148696
- Gene Aliases
- Chromosome Location
- Chr. 1: 207818379 - 207822596 on Build GRCh38
- UniGene ID
- Hs.125511
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026817.1 |
|
1 |
1689 |
|
GenBank mRNA |
AK125868.1 |
- |
1 |
1690 |
|
View Full Product Details