Gene Transcripts

Gene Symbol
FMO9P
Entrez Gene ID
116123
Gene Name
flavin containing monooxygenase 9 pseudogene
Gene Aliases
Chromosome Location
Chr. 1: 166603916 - 166625238 on Build GRCh38
UniGene ID
Hs.348539
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002925.2 4 340
GenBank mRNA BC014341.1 - 4 344


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CV028819.1 - 11 Chr. 1 - 166620817 - 166623791 [+]
GenBankmRNA BX283830.1 - 4 341 Chr. 1 - 166603916 - 166620871 [+]

View Full Product Details