Gene Transcripts

Gene Symbol
DKFZP434L187
Entrez Gene ID
26082
Gene Name
uncharacterized LOC26082
Gene Aliases
Chromosome Location
Chr. 15: 30196036 - 30214540 on Build GRCh38
UniGene ID
Hs.652128
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026771.1 3 1874
GenBank mRNA AL117445.1 - 2 1764
BC130408.1 - 3 1874
BC130410.1 - 3 1874

View Full Product Details