Gene Transcripts

Gene Symbol
ADAM5
Entrez Gene ID
255926
Gene Name
ADAM metallopeptidase domain 5 (pseudogene)
Gene Aliases
ADAM5P, TMDCII
Chromosome Location
Chr. 8: 39314663 - 39417226 on Build GRCh38
UniGene ID
Hs.369765
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_001448.1 4 243
GenBank mRNA AJ132821.1 - 4 243
AJ132822.1 - 4 243
AJ132823.1 - 4 243
AJ132824.1 - 3 189
AJ132825.1 - 2 109
BC047448.1 - 3 254
BC067864.1 - 3 220


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BI559754.1 - 4 278 Chr. 8 - 39314644 - 39346028 [+]
GenBankmRNA DB046693.1 - 4 270 Chr. 8 - 39314636 - 39325239 [+]
GenBankmRNA BI829318.1 - 3 196 Chr. 8 - 39314656 - 39375761 [+]
GenBankmRNA DB051731.1 - 4 274 Chr. 8 - 39314632 - 39325230 [+]
GenBankmRNA DB041573.1 - 3 202 Chr. 8 - 39314650 - 39345887 [+]
GenBankmRNA DB035078.1 - 4 274 Chr. 8 - 39314632 - 39324345 [+]
GenBankmRNA DB032119.1 - 2 155 Chr. 8 - 39314617 - 39345912 [+]
GenBankmRNA BU561620.1 - 4 273 Chr. 8 - 39314636 - 39345972 [+]
GenBankmRNA BU569030.1 - 3 225 Chr. 8 - 39314629 - 39346139 [+]
GenBankmRNA DB020111.1 - 4 274 Chr. 8 - 39314632 - 39345876 [+]
GenBankmRNA BX092331.1 - 3 196 Chr. 8 - 39314662 - 39335493 [+]
GenBankmRNA BG718414.1 - 4 319 Chr. 8 - 39314591 - 39345997 [+]
GenBankmRNA HY038535.1 - 4 265 Chr. 8 - 39314642 - 39324272 [+]
GenBankmRNA BI560028.1 - 3 224 Chr. 8 - 39314644 - 39375865 [+]
GenBankmRNA DB450693.1 - 3 221 Chr. 8 - 39314631 - 39324333 [+]
GenBankmRNA BQ441614.1 - 3 223 Chr. 8 - 39314632 - 39346136 [+]
GenBankmRNA DB029139.1 - 3 242 Chr. 8 - 39314610 - 39345877 [+]
GenBankmRNA DB459761.1 - 3 220 Chr. 8 - 39314632 - 39324292 [+]
GenBankmRNA DB032761.1 - 4 270 Chr. 8 - 39314636 - 39344508 [+]
GenBankmRNA DB044150.1 - 4 274 Chr. 8 - 39314632 - 39344100 [+]

View Full Product Details