Gene Transcripts

Gene Symbol
LINC00521
Entrez Gene ID
256369
Gene Name
long intergenic non-protein coding RNA 521
Gene Aliases
C14orf48, c14_5713
Chromosome Location
Chr. 14: 93997270 - 94011695 on Build GRCh38
UniGene ID
Hs.143845
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024182.1 2 78
NR_024183.1 2 330
NR_024184.1 2 330
GenBank mRNA AK097741.1 - 2 325
BC021728.2 - 2 329
BC031252.1 - 2 78
BI458986.1 - 2 334


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BI463117.1 - 2 83 Chr. 14 - 93997268 - 94001366 [+]

View Full Product Details