Gene Transcripts

Gene Symbol
TTTY6
Entrez Gene ID
84672
Gene Name
testis-specific transcript, Y-linked 6 (non-protein coding)
Gene Aliases
LINC00127, NCRNA00127, TTTY6A, TTY6
Chromosome Location
Chr. Y: 22439590 - 22441459 on Build GRCh38
UniGene ID
Hs.535085
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_001527.2 3 364

Gene Symbol
TTTY6B
Entrez Gene ID
441543
Gene Name
testis-specific transcript, Y-linked 6B (non-protein coding)
Gene Aliases
LINC00128, NCRNA00128, TTTY6, TTY6
Chromosome Location
Chr. Y: 22144987 - 22146834 on Build GRCh38
UniGene ID
Hs.535085
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002175.3 3 342

View Full Product Details