Gene Transcripts

Gene Symbol
SNHG9
Entrez Gene ID
735301
Gene Name
small nucleolar RNA host gene 9
Gene Aliases
NCRNA00062
Chromosome Location
Chr. 16: 1964996 - 1965504 on Build GRCh38
UniGene ID
Hs.115329
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003142.2 88
GenBank mRNA CR749453.1 - 89


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BI832564.1 - 1 51 Chr. 16 - 1965033 - 1965504 [+]
GenBankmRNA AW958849.1 - 104 Chr. - - [-]
GenBankmRNA AV762892.1 - 67 Chr. - - [-]
GenBankmRNA BU849390.1 - 1 58 Chr. 16 - 1965026 - 1965504 [+]
GenBankmRNA AA661893.1 - 1 71 Chr. 16 - 1965013 - 1965508 [+]
GenBankmRNA DW437557.1 - 35 Chr. - - [-]
GenBankmRNA BI964920.1 - 1 48 Chr. 16 - 1965036 - 1965504 [+]
GenBankmRNA CB110750.1 - 1 53 Chr. 16 - 1965031 - 1965508 [+]
GenBankmRNA DW441990.1 - 42 Chr. - - [-]
GenBankmRNA BF131329.1 - 59 Chr. - - [-]
GenBankmRNA BU960491.1 - 1 28 Chr. 16 - 1965059 - 1965504 [+]
GenBankmRNA CD174859.1 - 1 80 Chr. 16 - 1965028 - 1965504 [+]
GenBankmRNA BF217948.1 - 1 68 Chr. 16 - 1965016 - 1965497 [+]
GenBankmRNA BU565055.1 - 1 95 Chr. 16 - 1964992 - 1965504 [+]
GenBankmRNA DW442228.1 - 39 Chr. - - [-]
GenBankmRNA AA357636.1 - 1 105 Chr. 16 - 1964979 - 1965502 [+]
GenBankmRNA BC028232.1 - 1 50 Chr. 16 - 1965034 - 1965504 [+]
GenBankmRNA BX096205.1 - 1 124 Chr. 16 - 1964960 - 1965509 [+]
GenBankmRNA DW438767.1 - 1 74 Chr. 16 - 1965025 - 1965499 [+]
GenBankmRNA BU587988.1 - 28 Chr. - - [-]
GenBankmRNA BI962931.1 - 1 92 Chr. 16 - 1964992 - 1965504 [+]

View Full Product Details