Gene Transcripts

Gene Symbol
CEACAM22P
Entrez Gene ID
388550
Gene Name
carcinoembryonic antigen related cell adhesion molecule 22, pseudogene
Gene Aliases
Chromosome Location
Chr. 19: 44537058 - 44556917 on Build GRCh38
UniGene ID
Hs.446909
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027754.2 4 935
GenBank mRNA AK302997.1 - 5 861

View Full Product Details