Gene Transcripts
- Gene Symbol
- CEACAM22P
- Entrez Gene ID
- 388550
- Gene Name
- carcinoembryonic antigen related cell adhesion molecule 22, pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 19: 44537058 - 44556917 on Build GRCh38
- UniGene ID
- Hs.446909
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027754.2 |
|
5 |
1263 |
|
GenBank mRNA |
AK123850.1 |
- |
5 |
1263 |
|
AK302997.1 |
- |
6 |
1189 |
|
View Full Product Details