Gene Transcripts

Gene Symbol
FBXO22-AS1
Entrez Gene ID
692224
Gene Name
FBXO22 antisense RNA 1
Gene Aliases
FBXO22OS, FISTC1OS, HsT18082, NCRNA00053
Chromosome Location
Chr. 15: 75932683 - 75933130 on Build GRCh38
UniGene ID
Hs.591115
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003136.1 1 53
GenBank mRNA AA843761.1 - 1 57
AA931120.1 - 1 58
AI868357.1 - 1 57

View Full Product Details