Gene Transcripts
- Gene Symbol
- FBXO22-AS1
- Entrez Gene ID
- 692224
- Gene Name
- FBXO22 antisense RNA 1
- Gene Aliases
- FBXO22OS, FISTC1OS, HsT18082, NCRNA00053
- Chromosome Location
- Chr. 15: 75932683 - 75933130 on Build GRCh38
- UniGene ID
- Hs.591115
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003136.1 |
|
1 |
239 |
|
GenBank mRNA |
AA931120.1 |
- |
1 |
244 |
|
AI868357.1 |
- |
1 |
243 |
|
View Full Product Details