Gene Transcripts
- Gene Symbol
- GUCY2EP
- Entrez Gene ID
- 390226
- Gene Name
- guanylate cyclase 2E, pseudogene
- Gene Aliases
- GC-E, GCD, GUCY2E
- Chromosome Location
- Chr. 11: 76680166 - 76721789 on Build GRCh38
- UniGene ID
- Hs.440026
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024042.1 |
|
9 |
1150 |
|
View Full Product Details