Gene Transcripts
- Gene Symbol
- LOC283731
- Entrez Gene ID
- 283731
- Gene Name
- uncharacterized LOC283731
- Gene Aliases
- Chromosome Location
- Chr. 15: 74126373 - 74129278 on Build GRCh38
- UniGene ID
- Hs.631696
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027073.1 |
|
2 |
1728 |
|
GenBank mRNA |
AK095530.1 |
- |
2 |
1625 |
|
BC050067.1 |
- |
1 |
1628 |
|
View Full Product Details