Gene Transcripts

Gene Symbol
LOC202181
Entrez Gene ID
202181
Gene Name
SUMO interacting motifs containing 1 pseudogene
Gene Aliases
Chromosome Location
Chr. 5: 177618500 - 177672277 on Build GRCh38
UniGene ID
Hs.189914
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026921.1 2 347
GenBank mRNA Y13871.1 - 1 54

Gene Symbol
SIMC1
Entrez Gene ID
375484
Gene Name
SUMO interacting motifs containing 1
Gene Aliases
C5orf25, OOMA1, PLEIAD
Chromosome Location
Chr. 5: 176238359 - 176345991 on Build GRCh38
UniGene ID
Hs.719847
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308195.1 NP_001295124.1 2 371
NM_001308196.1 NP_001295125.1 4 579
XM_011534553.2 XP_011532855.1 3 4376
XM_011534554.2 XP_011532856.1 3 248
XM_011534556.2 XP_011532858.1 2 4245
XM_017009454.1 XP_016864943.1 2 179
XM_017009455.1 XP_016864944.1 3 489
XM_017009456.1 XP_016864945.1 3 385
GenBank mRNA AK126204.1 - 1 907
AK299336.1 - 2 331
BC037298.1 - 4 580 4823991
DC372557.1 - 2 331
HY028607.1 - 3 503
KF151170.1 - 2 239

View Full Product Details