Gene Transcripts

Gene Symbol
TMEM41B
Entrez Gene ID
440026
Gene Name
transmembrane protein 41B
Gene Aliases
Chromosome Location
Chr. 11: 9280654 - 9314768 on Build GRCh38
UniGene ID
Hs.594563
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015012.3 NP_055827.1 7 3746
NR_028491.2 8 1417
GenBank mRNA BC035034.1 - 7 3624
BC091524.1 - 8 1216 30512350

View Full Product Details