Gene Transcripts
- Gene Symbol
- RPL13AP17
- Entrez Gene ID
- 399670
- Gene Name
- ribosomal protein L13a pseudogene 17
- Gene Aliases
- RPL13A_6_816
- Chromosome Location
- Chr. 7: 78347242 - 78359458 on Build GRCh38
- UniGene ID
- Hs.568045
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003680.1 |
|
6 |
1408 |
|
GenBank mRNA |
BC027852.1 |
- |
6 |
1408 |
|
View Full Product Details