Gene Transcripts
- Gene Symbol
- SCARNA16
- Entrez Gene ID
- 677781
- Gene Name
- small Cajal body-specific RNA 16
- Gene Aliases
- ACA47
- Chromosome Location
- Chr. 17: 77089307 - 77089493 on Build GRCh38
- UniGene ID
- Hs.720923
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003013.1 |
|
1 |
121 |
|
GenBank mRNA |
AJ609486.1 |
- |
1 |
121 |
|
- Gene Symbol
- MIR6516
- Entrez Gene ID
- 102466864
- Gene Name
- microRNA 6516
- Gene Aliases
- hsa-mir-6516
- Chromosome Location
- Chr. 17: 77089417 - 77089497 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_106997.1 |
|
|
11 |
|
View Full Product Details