Gene Transcripts
- Gene Symbol
- ANKRD20A11P
- Entrez Gene ID
- 391267
- Gene Name
- ankyrin repeat domain 20 family member A11, pseudogene
- Gene Aliases
- C21orf81
- Chromosome Location
- Chr. 21: 13943769 - 13980444 on Build GRCh38
- UniGene ID
- Hs.364456
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027270.1 |
|
5, 6 |
1000 |
|
GenBank mRNA |
AF426257.1 |
- |
5, 6 |
1000 |
|
View Full Product Details