Gene Transcripts

Gene Symbol
TTTY14
Entrez Gene ID
83869
Gene Name
testis-specific transcript, Y-linked 14 (non-protein coding)
Gene Aliases
CYorf14, NCRNA00137, NCRNA00185, PRO2834, TTY14
Chromosome Location
Chr. Y: 18872501 - 19077547 on Build GRCh38
UniGene ID
Hs.138453
Species
Homo sapiens
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_125734.1
3 646
NR_125735.1
2 571
NR_125736.1
4 321
GenBank mRNA AF119903.1 - LIT2233
CYorf14
1 1088
BX280014.1 -
2 297
FY211103.1 -
2 234
FY211594.1 -
3 310
HY121109.1 -
4 314


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA HY130111.1 - 4 314 Chr. Y - 18872737 - 19075996 [-]
GenBankmRNA FY211412.1 - 2 148 Chr. Y - 18872765 - 19068798 [-]
GenBankmRNA BI823157.1 - 219 Chr. - - [-]

View Full Product Details