Gene Transcripts

Gene Symbol
BAIAP2-AS1
Entrez Gene ID
440465
Gene Name
BAIAP2 antisense RNA 1 (head to head)
Gene Aliases
Chromosome Location
Chr. 17: 81029133 - 81034719 on Build GRCh38
UniGene ID
Hs.448889
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026857.1 2 3642
GenBank mRNA AK027350.1 - 1 1506
AK056555.1 - 2 3624
AK075238.1 - 1 1605
BC110822.1 - 1 1651

View Full Product Details