Gene Transcripts

Gene Symbol
LRRC27
Entrez Gene ID
80313
Gene Name
leucine rich repeat containing 27
Gene Aliases
Chromosome Location
Chr. 10: 132330063 - 132381506 on Build GRCh38
UniGene ID
Hs.119897
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001309474.1 NP_001296403.1 1 128
XM_011540210.2 XP_011538512.1 1 126
GenBank mRNA DC352688.1 - 1 129


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BI771687.1 - 1 97 Chr. 10 - 132337138 - 132348118 [+]
GenBankmRNA DB499829.1 - 1 103 Chr. 10 - 132337132 - 132344577 [+]
GenBankmRNA DB493310.1 - 1 103 Chr. 10 - 132337132 - 132344569 [+]
GenBankmRNA BX436744.2 - 1 181 Chr. 10 - 132337054 - 132337904 [+]
GenBankmRNA DB485255.1 - 1 103 Chr. 10 - 132337132 - 132337698 [+]
GenBankmRNA DB505967.1 - 1 103 Chr. 10 - 132337132 - 132344584 [+]

View Full Product Details