Gene Transcripts

Gene Symbol
SPRR2C
Entrez Gene ID
6702
Gene Name
small proline rich protein 2C (pseudogene)
Gene Aliases
Chromosome Location
Chr. 1: 153140118 - 153141493 on Build GRCh38
UniGene ID
Hs.2421
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003062.1 2 101
GenBank mRNA BC120937.1 - 2 59
M21539.1 - 2 101

View Full Product Details