Gene Transcripts
- Gene Symbol
- TPTE2P3
- Entrez Gene ID
- 220115
- Gene Name
- transmembrane phosphoinositide 3-phosphatase and tensin homolog 2 pseudogene 3
- Gene Aliases
- TPTEps1
- Chromosome Location
- Chr. 13: 52488993 - 52587090 on Build GRCh38
- UniGene ID
- Hs.528448
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002793.1 |
|
24 |
2529 |
|
GenBank mRNA |
DQ577179.1 |
- |
1 |
21 |
|
- Gene Symbol
- TPTE2P2
- Entrez Gene ID
- 644623
- Gene Name
- transmembrane phosphoinositide 3-phosphatase and tensin homolog 2 pseudogene 2
- Gene Aliases
- Chromosome Location
- Chr. 13: 52218642 - 52323295 on Build GRCh38
- UniGene ID
- Hs.678973
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC041842.1 |
- |
7 |
3749 |
|
View Full Product Details