Gene Transcripts
- Gene Symbol
- TTTY16
- Entrez Gene ID
- 252948
- Gene Name
- testis-specific transcript, Y-linked 16 (non-protein coding)
- Gene Aliases
- NCRNA00139
- Chromosome Location
- Chr. Y: 7699357 - 7701247 on Build GRCh38
- UniGene ID
- Hs.522848
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001552.1 |
|
2 |
85 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AF527828.3 |
- |
2 |
85 |
|
Chr. Y - 7699357 - 7701247 [-] |
View Full Product Details