Gene Transcripts

Gene Symbol
TUBA3FP
Entrez Gene ID
113691
Gene Name
tubulin alpha 3f pseudogene
Gene Aliases
Chromosome Location
Chr. 22: 21008203 - 21014287 on Build GRCh38
UniGene ID
Hs.585006
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003608.1 1 469
GenBank mRNA AK057257.1 - 1 421
AK315686.1 - 1 417
BC009991.2 - 1 327
BI668929.1 - 1 478
CN368433.1 - 1 446

View Full Product Details