Gene Transcripts

Gene Symbol
BRD7P3
Entrez Gene ID
23629
Gene Name
bromodomain containing 7 pseudogene 3
Gene Aliases
BP75
Chromosome Location
Chr. 6: 118501373 - 118503846 on Build GRCh38
UniGene ID
Hs.648050
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002730.2 1 1823
GenBank mRNA BC033937.1 - 1 577


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BG714720.1 - 1 577 Chr. 6 - 118502621 - 118503354 [+]

View Full Product Details