Gene Transcripts
- Gene Symbol
- MSTO2P
- Entrez Gene ID
- 100129405
- Gene Name
- misato family member 2, pseudogene
- Gene Aliases
- MSTO2
- Chromosome Location
- Chr. 1: 155745768 - 155750688 on Build GRCh38
- UniGene ID
- Hs.656361
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC099909.1 |
- |
13 |
2324 |
|
- Gene Symbol
- MSTO1
- Entrez Gene ID
- 55154
- Gene Name
- misato 1, mitochondrial distribution and morphology regulator
- Gene Aliases
- LST005, MST
- Chromosome Location
- Chr. 1: 155610170 - 155614967 on Build GRCh38
- UniGene ID
- Hs.656547
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC070067.1 |
- |
10 |
3219 |
|
View Full Product Details