Gene Transcripts

Gene Symbol
SNHG6
Entrez Gene ID
641638
Gene Name
small nucleolar RNA host gene 6
Gene Aliases
HBII-276HG, NCRNA00058, U87HG
Chromosome Location
Chr. 8: 66921930 - 66925542 on Build GRCh38
UniGene ID
Hs.372680
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002599.1 1 38
GenBank mRNA AK057116.1 - 1 37
AY264285.1 - 1 38


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BI669107.1 - 1 93 Chr. 8 - 66921934 - 66925598 [-]
GenBankmRNA HY162808.1 - 1 37 Chr. 8 - 66921987 - 66925540 [-]
GenBankmRNA BM737762.1 - 1 25 Chr. 8 - 66921984 - 66925529 [-]
GenBankmRNA BQ924387.1 - 15 Chr. - - [-]
GenBankmRNA BI669733.1 - 1 42 Chr. 8 - 66921925 - 66925541 [-]
GenBankmRNA BI769050.1 - 20 Chr. - - [-]
GenBankmRNA DA126867.1 - 1 37 Chr. 8 - 66921984 - 66925541 [-]
GenBankmRNA BY796444.2 - 1 13 Chr. 8 - 66921935 - 66925517 [-]
GenBankmRNA BM016905.1 - 1 22 Chr. 8 - 66921935 - 66925526 [-]
GenBankmRNA BM737568.1 - 36 Chr. - - [-]
GenBankmRNA CB130641.1 - 1 37 Chr. 8 - 66922002 - 66925541 [-]
GenBankmRNA BU193261.1 - 1 35 Chr. 8 - 66921934 - 66925530 [-]
GenBankmRNA BU167862.1 - 1 38 Chr. 8 - 66921925 - 66925534 [-]
GenBankmRNA BU195948.1 - 1 20 Chr. 8 - 66921968 - 66925524 [-]
GenBankmRNA CN257859.1 - 1 15 Chr. 8 - 66921938 - 66925519 [-]
GenBankmRNA AJ712674.1 - 1 12 Chr. 8 - 66922010 - 66925516 [-]
GenBankmRNA BG034409.1 - 34 Chr. - - [-]
GenBankmRNA BM014460.1 - 1 21 Chr. 8 - 66921934 - 66925525 [-]
GenBankmRNA AA307876.1 - 1 21 Chr. 8 - 66922048 - 66925525 [-]
GenBankmRNA BM784351.1 - 1 37 Chr. 8 - 66921984 - 66925541 [-]
GenBankmRNA BF966290.2 - 1 42 Chr. 8 - 66921934 - 66925541 [-]
GenBankmRNA DB498305.1 - 1 88 Chr. 8 - 66921934 - 66925591 [-]
GenBankmRNA AW958356.1 - 22 Chr. - - [-]
GenBankmRNA BM768256.1 - 1 37 Chr. 8 - 66921968 - 66925541 [-]
GenBankmRNA AA429085.1 - 1 26 Chr. 8 - 66921958 - 66925527 [-]
GenBankmRNA BU596037.1 - 40 Chr. - - [-]
GenBankmRNA BQ440073.1 - 19 Chr. - - [-]
GenBankmRNA BF212415.1 - 1 42 Chr. 8 - 66921933 - 66925545 [-]
GenBankmRNA BM743937.1 - 1 36 Chr. 8 - 66921990 - 66925541 [-]
GenBankmRNA BI918140.1 - 23 Chr. - - [-]
GenBankmRNA HY294280.1 - 1 77 Chr. 8 - 66922000 - 66925542 [-]
GenBankmRNA DB471689.1 - 1 19 Chr. 8 - 66921984 - 66925523 [-]
GenBankmRNA HY117390.1 - 1 88 Chr. 8 - 66921984 - 66925591 [-]
GenBankmRNA BG034121.1 - 1 27 Chr. 8 - 66921938 - 66925531 [-]
GenBankmRNA BM842145.1 - 1 38 Chr. 8 - 66921984 - 66925542 [-]
GenBankmRNA BM842017.1 - 1 38 Chr. 8 - 66921984 - 66925542 [-]
GenBankmRNA HY279249.1 - 1 74 Chr. 8 - 66922022 - 66925541 [-]
GenBankmRNA BI550715.1 - 1 43 Chr. 8 - 66921934 - 66925542 [-]
GenBankmRNA BQ073523.1 - 15 Chr. - - [-]
GenBankmRNA HY066133.1 - 1 36 Chr. 8 - 66921948 - 66925540 [-]
GenBankmRNA BM741967.1 - 1 36 Chr. 8 - 66921991 - 66925541 [-]
GenBankmRNA CN482917.1 - 1 21 Chr. 8 - 66921934 - 66925525 [-]
GenBankmRNA BU565593.1 - 1 40 Chr. 8 - 66921984 - 66925541 [-]
GenBankmRNA AW630767.1 - 1 14 Chr. 8 - 66921971 - 66925518 [-]
GenBankmRNA HY112530.1 - 1 37 Chr. 8 - 66921948 - 66925540 [-]
GenBankmRNA HY075045.1 - 1 39 Chr. 8 - 66921984 - 66925540 [-]
GenBankmRNA DB569402.1 - 1 35 Chr. 8 - 66921936 - 66925539 [-]
GenBankmRNA CX752858.1 - 1 20 Chr. 8 - 66921934 - 66925524 [-]
GenBankmRNA DA301664.1 - 1 89 Chr. 8 - 66921910 - 66925593 [-]
GenBankmRNA DA154192.1 - 1 37 Chr. 8 - 66921818 - 66925541 [-]
GenBankmRNA HY084735.1 - 1 36 Chr. 8 - 66921987 - 66925540 [-]
GenBankmRNA BG703295.1 - 1 24 Chr. 8 - 66921984 - 66925525 [-]
GenBankmRNA BM737847.1 - 1 26 Chr. 8 - 66921984 - 66925530 [-]
GenBankmRNA CD513303.1 - 1 15 Chr. 8 - 66921984 - 66925519 [-]
GenBankmRNA BM840736.1 - 1 38 Chr. 8 - 66921996 - 66925542 [-]
GenBankmRNA BM015094.1 - 22 Chr. - - [-]
GenBankmRNA DB540433.1 - 1 19 Chr. 8 - 66921997 - 66925523 [-]
GenBankmRNA HY056578.1 - 1 36 Chr. 8 - 66921934 - 66925540 [-]
GenBankmRNA BM760648.1 - 1 38 Chr. 8 - 66921991 - 66925542 [-]
GenBankmRNA DA084810.1 - 1 38 Chr. 8 - 66921839 - 66925542 [-]
GenBankmRNA BM737712.1 - 1 23 Chr. 8 - 66921986 - 66925527 [-]
GenBankmRNA BU953849.1 - 1 63 Chr. 8 - 66921987 - 66925566 [-]
GenBankmRNA HY012009.1 - 1 36 Chr. 8 - 66921987 - 66925540 [-]
GenBankmRNA HY122223.1 - 1 36 Chr. 8 - 66921936 - 66925540 [-]

View Full Product Details