Gene Transcripts

Gene Symbol
GAS5
Entrez Gene ID
60674
Gene Name
growth arrest specific 5 (non-protein coding)
Gene Aliases
NCRNA00030, SNHG2
Chromosome Location
Chr. 1: 173863901 - 173867987 on Build GRCh38
UniGene ID
Hs.531856
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002578.2 12 605
GenBank mRNA BC038733.1 - 8 1673
BC070486.1 - 1 572


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CD174845.1 - 12 630 Chr. 1 - 173863900 - 173867988 [-]
GenBankmRNA BM011345.1 - 11 554 Chr. 1 - 173863901 - 173867018 [-]
GenBankmRNA CK820868.1 - 11 551 Chr. 1 - 173863904 - 173867017 [-]
GenBankmRNA BG331951.1 - 1 592 Chr. 6 - 89721170 - 89721816 [+]
GenBankmRNA BQ949316.1 - 1 604 Chr. 6 - 89721171 - 89721818 [+]
GenBankmRNA CD176412.1 - 1 623 Chr. 6 - 89721153 - 89721823 [+]
GenBankmRNA BG333353.1 - 600 Chr. - - [-]
GenBankmRNA BC003075.1 - 3427 Chr. - - [-]
GenBankmRNA CD173645.1 - 1 621 Chr. 6 - 89721151 - 89721826 [+]
GenBankmRNA CD248083.1 - 12 629 Chr. 1 - 173863901 - 173867989 [-]
GenBankmRNA BG470467.1 - 11 593 Chr. 1 - 173863900 - 173867047 [-]
GenBankmRNA BM781463.1 - 12 607 Chr. 1 - 173863908 - 173867989 [-]
GenBankmRNA BU181271.1 - 1 603 Chr. 6 - 89721170 - 89721815 [+]
GenBankmRNA BE787580.1 - 11 571 Chr. 1 - 173863908 - 173867038 [-]
GenBankmRNA CN280234.1 - 11 593 Chr. 1 - 173863904 - 173867047 [-]
GenBankmRNA BI260215.1 - 12 595 Chr. 1 - 173863901 - 173867977 [-]
GenBankmRNA BM046836.1 - 11 571 Chr. 1 - 173863901 - 173867036 [-]
GenBankmRNA BF314585.1 - 11 570 Chr. 1 - 173863901 - 173867036 [-]
GenBankmRNA CF127289.1 - 1 590 Chr. 6 - 89721172 - 89721819 [+]
GenBankmRNA BM047064.1 - 573 Chr. - - [-]
GenBankmRNA BU855344.1 - 1 478 Chr. 6 - 89721298 - 89721818 [+]
GenBankmRNA CB111494.1 - 12 606 Chr. 1 - 173863901 - 173867987 [-]
GenBankmRNA BG109420.1 - 10 512 Chr. 1 - 173863902 - 173866788 [-]
GenBankmRNA BQ184822.1 - 1 30 Chr. 1 - 173863253 - 173863941 [-]
GenBankmRNA CF130607.1 - 11 575 Chr. 1 - 173863901 - 173867035 [-]
GenBankmRNA BE277641.1 - 11 615 Chr. 1 - 173863907 - 173867054 [-]
GenBankmRNA CD385747.1 - 632 Chr. - - [-]
GenBankmRNA BG332918.1 - 1 593 Chr. 6 - 89721170 - 89726515 [+]
GenBankmRNA DR421845.1 - 1 598 Chr. 6 - 89721170 - 89721816 [+]
GenBankmRNA CD175280.1 - 12 627 Chr. 1 - 173863901 - 173867977 [-]
GenBankmRNA CD175024.1 - 12 630 Chr. 1 - 173863901 - 173867988 [-]
GenBankmRNA BF794520.1 - 1 591 Chr. 6 - 89721171 - 89721822 [+]
GenBankmRNA CD246672.1 - 12 633 Chr. 1 - 173863901 - 173867992 [-]
GenBankmRNA CN280231.1 - 11 592 Chr. 1 - 173863910 - 173867047 [-]
GenBankmRNA BM478043.1 - 1 591 Chr. 6 - 89721171 - 89721816 [+]
GenBankmRNA CD250276.1 - 12 624 Chr. 1 - 173863901 - 173867989 [-]
GenBankmRNA BM907339.1 - 1 618 Chr. 6 - 89721167 - 89721826 [+]
GenBankmRNA BM480323.1 - 12 604 Chr. 1 - 173863901 - 173867977 [-]
GenBankmRNA CN280235.1 - 1 592 Chr. 6 - 89721170 - 89721810 [+]
GenBankmRNA BQ214989.1 - 1 590 Chr. 6 - 89721172 - 89721830 [+]
GenBankmRNA BU859525.1 - 553 Chr. - - [-]
GenBankmRNA CD388871.1 - 12 626 Chr. 1 - 173863901 - 173867989 [-]
GenBankmRNA CD248920.1 - 1 631 Chr. 6 - 89721148 - 89721819 [+]
GenBankmRNA BF975855.1 - 11 574 Chr. 1 - 173863901 - 173867039 [-]
GenBankmRNA BI092136.1 - 11 560 Chr. 1 - 173863901 - 173867026 [-]
GenBankmRNA BG774686.1 - 1 593 Chr. 6 - 89721170 - 89721819 [+]
GenBankmRNA BQ934057.1 - 1 590 Chr. 6 - 89721171 - 89721822 [+]
GenBankmRNA BE901013.1 - 11 588 Chr. 1 - 173863906 - 173867047 [-]
GenBankmRNA CD555991.1 - 629 Chr. - - [-]
GenBankmRNA BF316352.1 - 588 Chr. - - [-]
GenBankmRNA BQ951229.1 - 11 569 Chr. 1 - 173863901 - 173867035 [-]
GenBankmRNA BQ951130.1 - 1 591 Chr. 6 - 89721171 - 89721819 [+]
GenBankmRNA BQ694556.1 - 11 581 Chr. 1 - 173863901 - 173867047 [-]
GenBankmRNA CN280223.1 - 11 564 Chr. 1 - 173863901 - 173867023 [-]
GenBankmRNA CD173104.1 - 12 625 Chr. 1 - 173863897 - 173867989 [-]
GenBankmRNA BM842238.1 - 12 607 Chr. 1 - 173863907 - 173867989 [-]
GenBankmRNA BM764699.1 - 12 606 Chr. 1 - 173863913 - 173867989 [-]
GenBankmRNA BM787205.1 - 12 607 Chr. 1 - 173863904 - 173867989 [-]
GenBankmRNA CD389474.1 - 629 Chr. - - [-]
GenBankmRNA BQ928231.1 - 11 570 Chr. 1 - 173863901 - 173867035 [-]
GenBankmRNA CN280232.1 - 11 592 Chr. 1 - 173863907 - 173867047 [-]
GenBankmRNA BU953699.1 - 374 Chr. - - [-]
GenBankmRNA CD249389.1 - 12 626 Chr. 1 - 173863901 - 173867990 [-]
GenBankmRNA BM770102.1 - 12 607 Chr. 1 - 173863909 - 173867989 [-]
GenBankmRNA CD176274.1 - 12 630 Chr. 1 - 173863901 - 173867989 [-]
GenBankmRNA BU540470.1 - 1 591 Chr. 6 - 89721171 - 89721822 [+]
GenBankmRNA CD177641.1 - 1 629 Chr. 6 - 89721153 - 89721823 [+]
GenBankmRNA BU163648.1 - 1 582 Chr. 6 - 89721180 - 89721818 [+]
GenBankmRNA BG028625.1 - 1 590 Chr. 6 - 89721172 - 89721813 [+]
GenBankmRNA CD386763.1 - 629 Chr. - - [-]
GenBankmRNA CD250191.1 - 12 629 Chr. 1 - 173863901 - 173867989 [-]
GenBankmRNA BQ228483.1 - 11 589 Chr. 1 - 173863901 - 173867049 [-]
GenBankmRNA BE543076.1 - 11 574 Chr. 1 - 173863907 - 173867038 [-]
GenBankmRNA CD246409.1 - 12 627 Chr. 1 - 173863901 - 173867989 [-]
GenBankmRNA CB151182.1 - 12 609 Chr. 1 - 173863914 - 173867990 [-]
GenBankmRNA BU566066.1 - 1 366 Chr. 6 - 89721411 - 89721830 [+]
GenBankmRNA CF129930.1 - 9 470 Chr. 1 - 173863900 - 173866549 [-]
GenBankmRNA CN280228.1 - 1 596 Chr. 6 - 89721172 - 89721810 [+]
GenBankmRNA BM739951.1 - 12 610 Chr. 1 - 173863911 - 173867993 [-]
GenBankmRNA BU500512.1 - 1 593 Chr. 6 - 89721174 - 89721819 [+]
GenBankmRNA BM764505.1 - 12 607 Chr. 1 - 173863905 - 173867989 [-]
GenBankmRNA AV758168.1 - 11 699 Chr. 1 - 173863902 - 173867056 [-]
GenBankmRNA CD173686.1 - 12 625 Chr. 1 - 173863901 - 173867990 [-]
GenBankmRNA CD249594.1 - 12 629 Chr. 1 - 173863901 - 173867989 [-]
GenBankmRNA CD248267.1 - 12 629 Chr. 1 - 173863901 - 173867989 [-]
GenBankmRNA CN280224.1 - 12 602 Chr. 1 - 173863901 - 173867977 [-]
GenBankmRNA BF794051.1 - 11 573 Chr. 1 - 173863901 - 173867037 [-]
GenBankmRNA BM764411.1 - 12 611 Chr. 1 - 173863905 - 173867989 [-]
GenBankmRNA BE391578.1 - 11 621 Chr. 1 - 173863903 - 173867047 [-]
GenBankmRNA CF129956.1 - 1 591 Chr. 6 - 89721171 - 89721817 [+]
GenBankmRNA CD387110.1 - 628 Chr. - - [-]
GenBankmRNA AV742993.1 - 1 512 Chr. 6 - 89721314 - 89721810 [+]
GenBankmRNA CD051158.1 - 624 Chr. - - [-]
GenBankmRNA CN280233.1 - 6 322 Chr. 1 - 173863901 - 173865489 [-]
GenBankmRNA BG259225.1 - 1 568 Chr. 6 - 89721191 - 89721820 [+]
GenBankmRNA CD249592.1 - 12 631 Chr. 1 - 173863901 - 173867989 [-]
GenBankmRNA CD175076.1 - 12 629 Chr. 1 - 173863901 - 173867989 [-]
GenBankmRNA BG177881.1 - 1 591 Chr. 6 - 89721170 - 89721826 [+]

View Full Product Details