Gene Transcripts
- Gene Symbol
- PWAR1
- Entrez Gene ID
- 145624
- Gene Name
- Prader Willi/Angelman region RNA 1
- Gene Aliases
- D15S227E, PAR-1, PAR1
- Chromosome Location
- Chr. 15: 25135642 - 25138053 on Build GRCh38
- UniGene ID
- Hs.612027
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
RNADB ID |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_022009.1 |
|
|
1 |
861 |
|
GenBank mRNA |
AF019616.1 |
- |
LIT2055 PAR1 |
1 |
864 |
|
View Full Product Details