Gene Transcripts
- Gene Symbol
- DLEU1
- Entrez Gene ID
- 10301
- Gene Name
- deleted in lymphocytic leukemia 1 (non-protein coding)
- Gene Aliases
- BCMS, BCMS1, DLB1, DLEU2, LEU1, LEU2, LINC00021, NCRNA00021, XTP6
- Chromosome Location
- Chr. 13: 50082169 - 50528643 on Build GRCh38
- UniGene ID
- Hs.591229
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
RNADB ID |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AJ412030.1 |
- |
LIT1922 BCMS |
1, 2 |
151 |
|
View Full Product Details